Documents
BR140005
BRAND® separating funnel with screw cap
size 250 mL, polypropylene
Synonym(s):
Funnels, Separating funnels
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
material
conical PTFE stopcock
polypropylene
packaging
pack of 1 ea
manufacturer/tradename
BRAND 140005
size
250 mL
General description
Polypropylene, high clarity for observation of contents. With polypropylene screw cap and special conical PTFE stopcock with retention device. Autoclavable (121 °C).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG