Key Documents
BR13906
BRAND® compact SILBERBRAND burette, with PTFE stopcock
volume 25 mL
Synonym(s):
Burettes, Compact burettes, SILBERBRAND
About This Item
UNSPSC Code:
41121808
NACRES:
NB.43
material
glass
manufacturer/tradename
BRAND 13906
volume measuring range
25 mL, accuracy: ±0.08 mL
volume
25 mL
General description
The New Generation of Burettes from BRAND. With PTFE stopcock, fast to dismantle and easy to clean. Simple repair - all individual components replaceable. Boro 3.3, removable PTFE stopcock. Error limits according to class B, DIN EN ISO 385. Calibrated to deliver (TD, Ex).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)