BMSD113201TP
Benchmark Zirconium Bulk Beads
bead diam. 0.1 mm, Molecular Biology (TriplePure)
Synonym(s):
grinding medium, grinding beads, beads for homogenization,BeadBug grinding beads
About This Item
Recommended Products
material
(zirconium)
grade
Molecular Biology (TriplePure)
packaging
pkg of 250 g
manufacturer/tradename
Benchmark Scientific D1132-01TP
bead diam.
0.1 mm
compatibility
BeadBug and BeadBlaster
Looking for similar products? Visit Product Comparison Guide
General description
Application
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service