BAF379440000
SP Bel-Art® Sterileware™ Plastic Mini Tongs
polystyrene, L 4.25 in. (11 cm), pkg of 25 ea (individually wrapped)
All Photos(3)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Recommended Products
material
polystyrene
packaging
pkg of 25 ea (individually wrapped)
manufacturer/tradename
Bel-Art F37944-0000
L
4.25 in. (11 cm)
General description
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals.
Features and Benefits
- Ridged jaws securely hold wet or dry samples
- One piece, sterile tongs are made from polystyrene plastic
- Individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)