Skip to Content
Merck
All Photos(3)

Documents

721379

Sigma-Aldrich

2-Aminophenylboronic acid hydrochloride

≥95%

Synonym(s):

2-Aminophenylboronic acid HCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C6H4B(OH)2 · HCl
CAS Number:
Molecular Weight:
173.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

solid

mp

145-156 °C

SMILES string

Cl.Nc1ccccc1B(O)O

InChI

1S/C6H8BNO2.ClH/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,9-10H,8H2;1H

InChI key

WPDASZCYRKGSTO-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of small molecules with a trihydroxybenzene motif as pan-selectin antagonists and potential antiinflammatory agents
  • Preparation of designed boronate ligands for glucose-selective holographic sensors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service