Skip to Content
Merck
All Photos(1)

Documents

647829

Sigma-Aldrich

PAMAM dendrimer, cystamine core, generation 2.0 solution

20 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2CH2CH2SSCH2CH2NH2]:(G=2);dendri PAMAM(NH2)16
Molecular Weight:
3348.36
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

amino surface groups

form

liquid

mol wt

generation 2

feature

core type cystamine core

concentration

20 wt. % in methanol

no. Surface Groups

16

bp

64.7 °C

density

0.860 g/mL at 25 °C

storage temp.

−20°C

Packaging

unit weight includes solvent

Legal Information

For research use only

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huang, B. H., Tomalia, D. A.
Journal of Luminescence, 111, 215-223 (2005)
DeMattei, C. R., Huang, B. H., Tomalia, D. A.
Nano Letters, 4, 771-777 (2004)
Anouk Zaal et al.
Frontiers in immunology, 11, 305-305 (2020-03-13)
Many tumors display alterations in the biosynthetic pathways of glycosylation, resulting in increased expression of specific tumor-associated glycan structures. Expression of these altered glycan structures is associated with metastasis and poor prognosis. Antigen presenting cells can recognize tumor-associated glycan structures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service