Skip to Content
Merck
All Photos(1)

Key Documents

522767

Sigma-Aldrich

(Aminomethyl)polystyrene

50-100 mesh, extent of labeling: 2.0 mmol/g loading, 1 % cross-linked

Synonym(s):

AM-polystyrene, Benzylamine polymer-bound, Poly(styrene-co-divinylbenzene), aminomethylated, Poly(styrene-co-vinylbenzylamine-co-divinylbenzene, Polystyrene crosslinked with divinylbenzene, aminomethylated

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

crosslinking

1 % cross-linked

reaction suitability

reaction type: Boc solid-phase peptide synthesis

extent of labeling

2.0 mmol/g loading

particle size

50-100 mesh

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Scavenges acids, acid chlorides, anhydrides, aldehydes and other electrophiles

Legal Information

Product of Agilent Technolgies

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service