Skip to Content
Merck
All Photos(1)

Key Documents

485268

Sigma-Aldrich

4-Oxo-TEMPO-d16, free radical

for ESR-spectroscopy, 97 atom % D

Synonym(s):

4-Oxo-,2,2,6,6-tetramethylpiperidine-d16-1-oxyl, TEMPONE-d16

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D16NO2
CAS Number:
Molecular Weight:
186.33
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

grade

for ESR-spectroscopy

isotopic purity

97 atom % D

Assay

99% (CP)

form

solid

mp

35 °C (lit.)

mass shift

M+16

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C1(N([O])C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])C(=O)C1([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C9H16NO2/c1-8(2)5-7(11)6-9(3,4)10(8)12/h5-6H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2

InChI key

WSGDRFHJFJRSFY-NMRSHPDNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service