Skip to Content
Merck
All Photos(1)

Documents

282677

Sigma-Aldrich

L-Lysine hydrate

97%

Synonym(s):

(S)-(+)-Lysine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)4CH(NH2)CO2H · xH2O
CAS Number:
Molecular Weight:
146.19 (anhydrous basis)
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

optical activity

[α]21/D +22°, c = 2 in 6 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

OC([C@@H](N)CCCCN)=O.O

InChI

1S/C6H14N2O2.H2O/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H2/t5-;/m0./s1

InChI key

HZRUTVAFDWTKGD-JEDNCBNOSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Hoon Han et al.
Nature communications, 5, 5633-5633 (2014-12-06)
The 26S proteasome is the primary machinery that degrades ubiquitin (Ub)-conjugated proteins, including many proteotoxic proteins implicated in neurodegeneraton. It has been suggested that the elevation of proteasomal activity is tolerable to cells and may be beneficial to prevent the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service