Skip to Content
Merck
All Photos(2)

Key Documents

219517

Sigma-Aldrich

Glycine sodium salt hydrate

98%, for peptide synthesis

Synonym(s):

Glycine monosodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CO2Na · xH2O
CAS Number:
Molecular Weight:
97.05 (anhydrous basis)
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

Glycine sodium salt hydrate, 98%

Quality Level

Assay

98%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

197-201 °C (lit.)

application(s)

peptide synthesis

SMILES string

[Na+].[H]O[H].NCC([O-])=O

InChI

1S/C2H5NO2.Na.H2O/c3-1-2(4)5;;/h1,3H2,(H,4,5);;1H2/q;+1;/p-1

InChI key

CIJQGPVMMRXSQW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service