Skip to Content
Merck
All Photos(1)

Key Documents

196371

Sigma-Aldrich

4-Bromo-2,6-xylenol

97%

Synonym(s):

4-Bromo-2,6-dimethylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(CH3)2OH
CAS Number:
Molecular Weight:
201.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

74-78 °C (lit.)

SMILES string

Cc1cc(Br)cc(C)c1O

InChI

1S/C8H9BrO/c1-5-3-7(9)4-6(2)8(5)10/h3-4,10H,1-2H3

InChI key

ZLVFYUORUHNMBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iodine initiated polymerization of 4-bromo-2,6-xylenol in tetrahydrofuran solution has been investigated. 4-Bromo-2,6-xylenol on bromination in chloroform or carbobn tetrachloride yields 3,4-dibromo-2,6-xylenol and 3,4,5-tribromo-2,6-xylenol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymerization of Three Bromo-2, 6-xylenols in THF Solution.
Semsarzadeh MA and Price C.
Macromolecules, 10(2), 482-485 (1977)
Spontaneous rearrangement of o-quinobromides.
Ershov VV and Volod'kin AA.
Russian Chemical Bulletin, 14(2), 313-318 (1965)
Yang Wu et al.
Water research, 169, 115202-115202 (2019-11-02)
The degradation of tetrabromobisphenol A (TBBPA), one of the most widely used brominated flame retardant, was evaluated during peroxymonosulfate (PMS) oxidation. TBBPA degradation was pH-dependent, with peak degradation rate constants occurring at pH 8.0-9.0, which was distinct from some other
Nikola Vaňková et al.
Journal of chromatography. A, 1409, 152-158 (2015-07-29)
The effect of gradient steepness on the kinetic performance limits and peak compression effects has been assessed in gradient mode for the separation of phenol derivatives using columns packed with 2.6μm core-shell particles. The effect of mobile-phase velocity on peak

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service