Merck
  • A matrix metalloproteinase 1-cleavable composite peptide derived from transforming growth factor β-inducible gene h3 potently inhibits collagen-induced arthritis.
A matrix metalloproteinase 1-cleavable composite peptide derived from transforming growth factor β-inducible gene h3 potently inhibits collagen-induced arthritis.
Arthritis and rheumatism (2013-03-20)
Eon Jeong Nam, Jin Hee Kang, Shijin Sung, Keum Hee Sa, Kyung Hoon Kim, Jae Seok Seo, Jong-Ho Kim, Seung Woo Han, In San Kim, Young Mo Kang
RESUMEN
Transforming growth factor β-inducible gene h3 (βIG-H3), which is abundantly expressed in rheumatoid synovium, and matrix metalloproteinases (MMP) play important roles in the pathogenesis of rheumatoid arthritis (RA). The aim of this study was to determine the therapeutic efficacy of βIG-H3-derived peptides using MMP-1-dependent target tissue delivery in chronic inflammatory arthritis. Peptides developed from βIG-H3 derivatives, including the second and fourth YH peptides, the fourth fas-1 domain, the fourth fas-1 domain truncated for H1 and H2 sequences (dhfas-1), and an MMP-1- cleavable composite peptide (MFK24), were cloned. We confirmed the specificity of MFK24 cleavage by immunoblot analysis after treatment with different proteases. The YH18 peptide in the fourth fas-1 domain of βIG-H3 was weakly effective in suppressing arthritis severity in mice with collagen-induced arthritis (CIA). Treatment with higher-dose dhfas-1 (30 mg/kg) showed remarkable efficacy, whereas treatment with a lower dose (10 mg/kg) resulted in only partial improvement. MFK24, a composite peptide consisting of dhfas-1 and RGD peptide linked by MMP-1 substrate, was cleaved specifically by MMP-1. The adhesion and migration of NIH3T3 cells mediated by βIG-H3 were inhibited by MFK24 at a low concentration. MFK24 suppressed the adhesion of NIH3T3 cells more efficiently compared with murine dhfas-1 (MFK00) or RGD, either alone or in combination. The therapeutic efficacy of MFK24 in mice with CIA was remarkably enhanced, with consistently reduced expression of inflammatory mediators within joint tissue. This proof-of-concept study showed that an MMP-cleavable composite peptide, based on βIG-H3 derivatives, had markedly improved therapeutic efficacy in chronic inflammatory arthritis, implicating a new expandable strategy for enhancement of the efficacy of 2 different active molecules in RA.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Arg-Gly-Asp, ≥97% (TLC)
Sigma-Aldrich
Anti-MMP-1 Antibody, a.a. 332-350 hMMP1, clone 41-1E5, clone 41-1E5, Chemicon®, from mouse