Merck
DNA nanostructure serum stability: greater than the sum of its parts.
Chemical communications (Cambridge, England) (2013-01-05)
Justin W Conway, Christopher K McLaughlin, Katherine J Castor, Hanadi Sleiman
RESUMEN
Simple chemical modifications to oligonucleotide ends with hexaethylene glycol and hexanediol are shown to significantly increase nuclease resistance under serum conditions. The modified oligonucleotides were used to construct DNA prismatic cages in a single step and in quantitative yield. These cages further stabilize their strands towards nucleases, with lifetimes of 62 hours in serum. The cages contain a large number of single-stranded regions for functionalization, illustrating their versatility for biological applications.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hexaethylene glycol, 97%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad