Documentos clave
57351
Supelco
Replacement Ferrules for SPME/HPLC Interface, pk 10
About This Item
Código UNSPSC:
12000000
envase
pkg of 10 ea
técnicas
solid phase microextraction (SPME): suitable
aplicaciones
food and beverages
compatibilidad
for use with SPME/HPLC interface
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
José Carlos Rodrigues et al.
Journal of chromatography. A, 1105(1-2), 208-212 (2005-12-28)
The aim of the study described in this report has been the development and the evaluation of a new improved interface to be operated under continuous heating, for on-line coupling solid-phase microextraction (SPME) to high-performance liquid chromatography (HPLC). Heating is
Cong Yao et al.
Journal of chromatography. A, 1216(6), 948-955 (2008-12-26)
A solid-phase microextraction (SPME)-high-performance liquid chromatography (HPLC) approach is used, for the first time, to study the partitioning behavior of eight aromatic analytes to three imidazolium-based ionic liquid micelles, namely, 1-hexadecyl-3-methylimidazolium bromide (HDMIm-Br), 1-hexadecyl-3-butylimidazolium bromide (HDBIm-Br), and 1,3-didodecylimidazolium bromide (DDDDIm-Br).
Effect of Heating the Interface on Chromatographic Performance of SPME Coupled to HPLC
Daimon, H., et al.
Analytical Communications, 34(11), 365-369 (1997)
SPME-LC Determination of Fluoxetine and Norfluoxetine in Plasma Using a Heated Liquid Flow Through Interface
Fernandes, C., et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 847(2), 217-233 (2007)
Effect of Heating the Interface on Chromatographic Performance of SPME Coupled to HPLC
Daimon, H., et al.
Analyst, 122(11), 365-369 (1997)