Documentos clave
Z762814
Eppendorf® Combitips advanced® pipette tips, Biopur® grade
maximum volume 25 mL
Sinónimos:
pipetting tips
About This Item
Código UNSPSC:
41121600
NACRES:
NB.01
Materiales
red tip
esterilidad
sterile
Características
barrier: no
envase
pack of 100 ea individually wrapped
fabricante / nombre comercial
Eppendorf® 0030089685
volumen máximo
25 mL
Descripción general
The Combitips advanced are optimized for all Repeater models to create the perfect connection!
The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.
The "Biopur" tips are individually blister-wrapped and feature an access tab which makes them easier to open, even with gloves.
Características y beneficios
  • pyrogen-free
  • free of human DNA
  • free of bacterial DNA
  • RNase-free
  • PCR inhibitor-free
  • DNase-free
  • ATP-free
Información legal
Biopur is a registered trademark of Eppendorf AG
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)