Documentos clave
Z659533
Bandelin SONOREX Ultrasonic bath accessories
tank lid to fit Z659517 and Z659509
About This Item
Código UNSPSC:
41103706
NACRES:
NB.74
Materiales
stainless steel
fabricante / nombre comercial
Bandelin D 52
disponibilidad
not available in USA and Canada
Descripción general
Stainless steel lid for an ultrasonic bath protects the bath liquid from external contamination and prevents unpleasant odors from the cleaning or disinfection agents added. In addition, the noise emitted when the ultrasonic cleaner is in operation, which may be perceived as annoying, is reduced. The lids are designed in such a way that any condensation that occurs is directed back into the bath.
Características y beneficios
  • protection of the bath fluid from contamination
  • sound-absorbing
  • shortens the heating phase
Información legal
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)