Z646725
BRAND® Transferpette® S benchtop mount
holds up to 6 single or multichannel Transferpette S Pipettes
About This Item
Código UNSPSC:
41122801
NACRES:
NB.01
Productos recomendados
Compatibilidad de esterilización
autoclavable compatible
Características
Holds 6 x pipetors
fabricante / nombre comercial
BRAND 704805
L × An × Al
13.125 in. × 8 in. × 7 in.
Descripción general
Freely rotatable carousel stand provides a convenient holding area and ready access for up to six pipettors at a time.
- Holds up to 6 single or multichannel Transferpette S Pipettes
- Dimensions: L 13.125" x W 8" x H 7"
- Fully autoclavable: 121°C (250°F)
Otras notas
BR704805 (India)
Información legal
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)