Z635049
Biohit Proline® single channel electronic pipettor
volume 10-500 μL
descripción
without charger
fabricante / nombre comercial
Sartorius 710100
disponibilidad
not available in ROW, USA and Canada
volumen
10-500 μL
Descripción general
Sartorius Proline electronic pipettors of the Biohitfamily offer effortless and rapid pipetting with high levels of accuracy and precision. They offer a wide variety of pipetting operations, making the Proline range the complete solution for all pipetting needs. Proline pipettors are ergonomically designed and the tip ejection is easily operated with four fingers to avoid strain and fatigue during pipetting. A microprocessor-controlled piston ensures optimal precision and accuracy at the same time being user-independent, delivering reliable results consistently. Proline liquid handling operations include:
- Forward Pipetting
- Reverse Pipetting (excl. 1200, 5000)
- Multiple-Dispensing
- Multi-Aspirating (only 1200)
- Diluting (excl. mcp 1200)
- Sequential-Dispensing (only 1200, 5000)
- Post-Delivery Mixing
Información legal
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)