Documentos
Z601209
Hettich® MIKRO 120 centrifuge accessories
adapter for 0.4 mL tubes 6x45 mm
About This Item
Código UNSPSC:
41103911
NACRES:
NB.22
Características
Holds 0.2 mL tubes
envase
pack of 6 ea
fabricante / nombre comercial
Hettich 2024
técnicas
centrifugation: suitable
compatibilidad
for use with 1213-A, 1226-A, 1252-A, 1258-A, 2428, 2434, 2437 (rotors)
Información legal
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG