Documentos
RF55165-EP
Rotor for Eppendorf® MiniSpin® / MiniSpin® Plus
F-55-16-5-PCR, non aerosol tight, to hold, 2 x 8-tube PCR strips, lid
About This Item
Código UNSPSC:
41103909
Características
lid
to hold 2 x 8-tube PCR strips
fixed angle rotor
non aerosol tight
envase
pack of 1 ea
fabricante / nombre comercial
Eppendorf® 022665821 (NA)
Eppendorf® 5452727007 (EU)
Parámetros
14,100 × g max. RCF
técnicas
PCR: suitable
centrifugation: suitable
compatibilidad
for use with centrifuge models MiniSpin/MiniSpin Plus
Descripción general
Rotors for use with Eppendorf MiniSpin and MiniSpin Plus. Rotors are fixed angle and include quick-Lock lids.
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Minispin is a registered trademark of Eppendorf AG