EPU9043-0006
CO2 Back-up System
for Eppendorf® CryoCube® F570 series, Premium, and HEF® freezers, AC/DC input 115 V / 208 V, 50/60 Hz, US 3-pin plug
About This Item
Código UNSPSC:
41121800
Productos recomendados
fabricante / nombre comercial
Eppendorf® U9043-0006
Categorías relacionadas
Descripción general
Loss of power is no problem with back-up systems, which are designed to maintain the contents of your freezer at a preset temperature should a power outage arise.
Características y beneficios
Equipped with a battery back-up to temporarily protect the contents of your freezer in case of power failure
Available in either liquid CO2 and LN2. Liquid CO2 can maintain temperatures between -50 C and -70 C, where as LN2 can go down to -85 C
Both systems can be factory installed with your freezer order or retrofitted on location
Info: CO2 and LN2 are not interchangeable systems
Available in either liquid CO2 and LN2. Liquid CO2 can maintain temperatures between -50 C and -70 C, where as LN2 can go down to -85 C
Both systems can be factory installed with your freezer order or retrofitted on location
Info: CO2 and LN2 are not interchangeable systems
Información legal
CryoCube is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
HEF is a registered trademark of Eppendorf, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)