EP6731011345
Eppendorf® CellXpert® C170i CO2 Incubator
Oxygen control, water/humidity monitoring, inner door with 4 door segments, handle right side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
About This Item
Código UNSPSC:
41104405
envase
pack of 1 ea
fabricante / nombre comercial
Eppendorf® 6731011345
Categorías relacionadas
Descripción general
CellXpert® C170i, inner door with 4 door segments, handle right side, 1 – 20 % oxygen control, water and humidity monitoring package, 100 – 127 V/50 – 60 Hz (US/JP/South America)
Características y beneficios
- water and humidity monitoring package
- 1 – 20 % oxygen control
- handle right side
- inner door with 4 door segments
- 180° HTD (standard with C170i)
- including water tray and four shelves (stainless steel)
- 100 – 127 V, 50 – 60 Hz
Información legal
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)