Documentos clave
EP5810747007
Sigma-Aldrich
Eppendorf® Adapter for 500mL rectangular bucket
for use with 2,6-7 mL Vacutainer
About This Item
Código UNSPSC:
41121815
NACRES:
NB.02
envase
pack of 2 ea
fabricante / nombre comercial
Eppendorf® 5810747007
técnicas
centrifugation: suitable
compatibilidad
for use with 2,6-7 mL Vacutainer
Descripción general
Holds 18 round-bottom tubes 2.6 – 7 mL and can also hold 500 mL rectangular beakers. To be used in Rotor A-4-81 and Rotor S-4x500. Supplied as 2 EA.
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)