Key Documents
EP4920602008
Eppendorf® Reference 2 lower part
single-channel, lower part with separate serial number, medium gray, size 0.5-10 μL
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Características
single-channel
lower part with separate serial number
envase
pkg of 1 ea
fabricante / nombre comercial
Eppendorf® 4920602008
tamaño
0.5-10 μL
color
medium gray
Descripción general
The name, Reference, stands for extraordinary precision and accuracy, a long service lifes and an ergonomic design. The new Reference 2 boasts these proven premium characteristics with its state-of-the-art technolgy, making it a reliable partner for you and your work. The time saving single-button operation is unrivalled in the new multichannel versions.
Información legal
Eppendorf is a registered trademark of Eppendorf AG