Documentos clave
CLS139532HTC
Corning® high-temp PBT screw cap
For high temps, GL 32 solid top, with PTFE faced silicone liner
About This Item
Código UNSPSC:
41121800
NACRES:
NB.11
Materiales
PTFE liner
red PBT cap
esterilidad
non-sterile
envase
pack of 10
Adaptador
GL 32 solid top
Descripción general
The red Corning polybutylene terephthalate (PBT) high temperature caps have a GL32 thread, can be dry heat sterilized at up to 180 deg. C and have a Polytetrafluorethylene (PTFE) liner for efficient sealing.
Información legal
Corning is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)