Documentos clave
64013BC
Millipore
MVP ICON® Field Care Kit
BioControl®, for use with MVP ICON® instruments
About This Item
Código UNSPSC:
41122409
Nivel de calidad
Línea del producto
MVP ICON®
fabricante / nombre comercial
MVP ICON®
aplicaciones
food and beverages
compatibilidad
MVP ICON®
Descripción general
MVP ICON® Sample Chamber Field Care Kit (10 Dry Cleaning Swabs)
Aplicación
MVP ICON® field care kit used for cleaning and decontamination of the internal sample chamber area. The MVP ICON® instrument is used for cleaning verification to monitor adherence to both sanitation and HACCP programs. Usage in some environments may introduce liquids, residue, and other contaminants into the sample chamber area, which can potentially affect performance including causing inaccurate ATP readings. This field care kit allows users to clean and decontaminate the internal sample chamber area of the instrument on-site.
Envase
Clear re-sealable plastic bag
Información legal
BioControl is a registered trademark of BioControl Systems, Inc.
MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)