Saltar a
MASVFCASE
Portable case
For use with MAS-100 VF®
Iniciar sesión para ver los precios por organización y contrato.
Seleccione un Tamaño
Cambiar Vistas
A ustedes/SKUDisponibilidadPrecio
1 unit
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
443,00 €
Acerca de este artículo
NACRES:
JA.11
UNSPSC Code:
41103403
Feature:
portable
443,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Solicitar más información
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarle
feature
portable
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with MAS-100 VF®
General description
The Portable case is an additional accessory for MAS-100 VF®, used for safe transport of the equipment. The carrying case provides space for one MAS-100 VF® air sampler including perforated lid and dust cover, USB memory stick, allen key, battery charger and documentation sheets such as Feller table and calibration certificates.
Application
The Portable case is used for safe carrying of MAS-100 VF® sampler that finds application in air sampling in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.
Legal Information
MAS-100 VF is a registered trademark of MBV AG
Comparar elementos similares
Ver comparación completa
Mostrar Diferencias
1 of 1
Este artículo
1.441501.173341.73020
Case
Supelco
1.73020
Case
description
For use with MAS-100 VF®
description
for use with RCS® High Flow Touch
description
for use with MAS-100 Atmos®
description
-
feature
portable
feature
portable
feature
portable
feature
-
compatibility
for use with MAS-100 VF®
compatibility
for use with RCS® High Flow Touch
compatibility
for use with MAS-100 Atmos®
compatibility
for use with Spectroquant® Prove 100, for use with Spectroquant® Prove 300, for use with Spectroquant® Prove 600
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
application(s)
compressed gas monitoring
food and beverages
pharmaceutical
application(s)
-
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)