HTS065M
ChemiSCREEN Membrane Preparation Recombinant Human MT1 Melatonin Receptor
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
About This Item
descripción
GPCR Class A
protein target MT1
Nivel de calidad
técnicas
radioligand binding assay (RLBA): suitable
Nº de acceso UniProt
Información sobre el gen
human ... MTNR1A(4543)
Descripción general
FUNCTION: SwissProt: P48039 # High affinity receptor for melatonin. Likely to mediates the reproductive and circadian actions of melatonin. The activity of this receptor is mediated by pertussis toxin sensitive G proteins that inhibit adenylate cyclase activity.
SIZE: 350 amino acids; 39375 Da
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
TISSUE SPECIFICITY: Expressed in hypophyseal pars tuberalis and hypothalamic suprachiasmatic nuclei (SCN). Hippocampus.
SIMILARITY: SwissProt: P48039 ## Belongs to the G-protein coupled receptor 1 family.
SIZE: 350 amino acids; 39375 Da
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
TISSUE SPECIFICITY: Expressed in hypophyseal pars tuberalis and hypothalamic suprachiasmatic nuclei (SCN). Hippocampus.
SIMILARITY: SwissProt: P48039 ## Belongs to the G-protein coupled receptor 1 family.
Aplicación
Radioligand binding assay and GTPγS binding.
Forma física
Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membranes protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.
Packaging method: Membranes protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.
Almacenamiento y estabilidad
Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.
Nota de análisis
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 500mM NaCl. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, filtered and stored at 4°C
Radioligand: [125I] Iodomelatonin (Perkin Elmer#: NEX-236)
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 15-fold signal:background with 125I-labeled Iodomelatonin at 0.5 nM
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, filtered and stored at 4°C
Radioligand: [125I] Iodomelatonin (Perkin Elmer#: NEX-236)
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 15-fold signal:background with 125I-labeled Iodomelatonin at 0.5 nM
Cláusula de descargo de responsabilidad
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificados de análisis (COA)
Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»
¿Ya tiene este producto?
Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.
Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.
Póngase en contacto con el Servicio técnico