Z407674
Quartz wall clock-thermometer-hygrometer
About This Item
Código UNSPSC:
41122411
Descripción general
Useful for the laboratory, office, or computer room. Measures RH from 0 to 100% in 2% subdivisions and temperature in °F. Attractive 93/4 in. diameter black face with fluorescent numerals makes it an easy-to-read addition to your office. Requires one AA battery (not included).
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)