Merck
  • Exclusion of alternative exon 33 of CaV1.2 calcium channels in heart is proarrhythmogenic.
Exclusion of alternative exon 33 of CaV1.2 calcium channels in heart is proarrhythmogenic.
Proceedings of the National Academy of Sciences of the United States of America (2017-05-12)
Guang Li, Juejin Wang, Ping Liao, Peter Bartels, Hengyu Zhang, Dejie Yu, Mui Cheng Liang, Kian Keong Poh, Chye Yun Yu, Fengli Jiang, Tan Fong Yong, Yuk Peng Wong, Zhenyu Hu, Hua Huang, Guangqin Zhang, Mary Joyce Galupo, Jin-Song Bian, Sathivel Ponniah, Scott Lee Trasti, Kelvin See, Roger Foo, Uta C Hoppe, Stefan Herzig, Tuck Wah Soong
ABSTRACT
Alternative splicing changes the CaV1.2 calcium channel electrophysiological property, but the in vivo significance of such altered channel function is lacking. Structure-function studies of heterologously expressed CaV1.2 channels could not recapitulate channel function in the native milieu of the cardiomyocyte. To address this gap in knowledge, we investigated the role of alternative exon 33 of the CaV1.2 calcium channel in heart function. Exclusion of exon 33 in CaV1.2 channels has been reported to shift the activation potential -10.4 mV to the hyperpolarized direction, and increased expression of CaV1.2Δ33 channels was observed in rat myocardial infarcted hearts. However, how a change in CaV1.2 channel electrophysiological property, due to alternative splicing, might affect cardiac function in vivo is unknown. To address these questions, we generated mCacna1c exon 33-/--null mice. These mice contained CaV1.2Δ33 channels with a gain-of-function that included conduction of larger currents that reflects a shift in voltage dependence and a modest increase in single-channel open probability. This altered channel property underscored the development of ventricular arrhythmia, which is reflected in significantly more deaths of exon 33-/- mice from β-adrenergic stimulation. In vivo telemetric recordings also confirmed increased frequencies in premature ventricular contractions, tachycardia, and lengthened QT interval. Taken together, the significant decrease or absence of exon 33-containing CaV1.2 channels is potentially proarrhythmic in the heart. Of clinical relevance, human ischemic and dilated cardiomyopathy hearts showed increased inclusion of exon 33. However, the possible role that inclusion of exon 33 in CaV1.2 channels may play in the pathogenesis of human heart failure remains unclear.
Sigma-Aldrich
Screen cup for CD-1, size 85 mL
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid