Merck
  • Protocol to downregulate GABAergic-astrocyte signaling via astrocyte-selective ablation of GABAB receptor in adult mice.
Protocol to downregulate GABAergic-astrocyte signaling via astrocyte-selective ablation of GABAB receptor in adult mice.
STAR protocols (2022-09-15)
Sara Mederos, Andrea Sánchez-Ruiz, Gertrudis Perea
ABSTRACT
Here, we present a protocol to selectively downregulate GABAB receptor (GABABR) expression in astrocytes of mouse medial prefrontal cortex (mPFC). We first describe the procedure of surgeries and viral injections. We then detail genetic, histological, and functional characterizations of astrocytic GABABR ablation using RT-PCR, imaging, and behavioral assays. The use of GABAB flox mice can be easily adapted to generate astrocyte-selective GABABR ablation in different brain areas and postnatal stages, leading to local downregulation of GABAergic-astrocyte signaling without developmental issues. For complete details on the use and execution of this protocol, please refer to Mederos et al. (2021).
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-GABA B Receptor R1 Antibody, serum, from guinea pig
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse