Skip to Content
Merck
All Photos(1)

Key Documents

V900790

Sigma-Aldrich

3,5-Dihydroxybenzoic acid

Vetec, reagent grade, 97%

Synonym(s):

α-Resorcylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
2207864
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

97%

mp

236-238 °C (dec.) (lit.)

SMILES string

OC(=O)c1cc(O)cc(O)c1

InChI

1S/C7H6O4/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3,8-9H,(H,10,11)

InChI key

UYEMGAFJOZZIFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

200 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matti Marklund et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(11-12), 888-894 (2010-03-13)
Alkylresorcinols (ARs) are phenolic lipids present at high concentrations in the outer parts of rye and wheat kernels and have been proposed as biomarkers for intake of whole grain and bran products of these cereals. AR are absorbed in the
Qian Cao et al.
Biosensors & bioelectronics, 25(12), 2680-2685 (2010-06-01)
A colorimetric, label-free, and nonaggregation-based gold nanoparticles probe has been developed for the detection of melamine. Gold nanoparticles were generated using 3,5-dihydroxybenzoic acid as a reducer without adding gold nanoparticle seeds at room temperature. Upon the addition of melamine, the
Y Matsumoto et al.
Journal of pharmaceutical sciences, 81(5), 428-431 (1992-05-01)
To reveal the role of the band 3 anion transport protein of the erythrocyte membrane in drug transport through the membrane, the possible effects of inhibitors of anion transport on the permeability of some anionic drugs were examined. The amounts
Anja Koskela et al.
Journal of agricultural and food chemistry, 56(17), 7678-7681 (2008-08-12)
This study presents the optimization and validation of a rapid protocol for quantifying alkyresorcinol (AR) metabolites 3,5-dihydroxybenzoic acid (DHBA) and 3-(3,5-dihydroxyphenyl)-1-propanoic acid (DHPPA) in plasma, using high-performance liquid chromatography (HPLC) coupled with a coulometric electrode array detector. Syringic acid (SyrA)
Silke C Wenzel et al.
Chembiochem : a European journal of chemical biology, 9(16), 2711-2721 (2008-10-31)
Kendomycin is a bioactive polyketide that is produced by various Streptomyces strains. It displays strong antibiotic activities against a wide range of bacteria and exhibits remarkable cytotoxic effects on the growth of several human cancer cell lines. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service