Skip to Content
Merck
All Photos(1)

Key Documents

V900778

Sigma-Aldrich

Propiophenone

Vetec, reagent grade, 99%

Synonym(s):

Ethyl phenyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COC2H5
CAS Number:
Molecular Weight:
134.18
Beilstein:
606215
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor pressure

1 mmHg ( 50 °C)

product line

Vetec

Assay

99%

refractive index

n20/D 1.526 (lit.)

bp

218 °C (lit.)

mp

17-19 °C (lit.)

density

1.009 g/mL at 20 °C
1.009 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)c1ccccc1

InChI

1S/C9H10O/c1-2-9(10)8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

KRIOVPPHQSLHCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Tang et al.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 21(3), 236-241 (2015-02-07)
Nontuberculous mycobacteria infection is a growing global concern, but data from Asia are limited. This study aimed to describe the distribution and antibiotic susceptibility profiles of rapidly growing mycobacterium (RGM) isolates in Singapore. Clinical RGM isolates with antibiotic susceptibility tests
Ruben De Pauw et al.
Journal of chromatography. A, 1420, 129-134 (2015-10-20)
A new system is proposed for applying fast temperature gradients in liquid chromatography. It consists of a 0.7 mm × 150 mm fused-silica column coated with a 50 μm Nickel-layer, which is connecting with a power source and a temperature
Nikola Vaňková et al.
Journal of chromatography. A, 1409, 152-158 (2015-07-29)
The effect of gradient steepness on the kinetic performance limits and peak compression effects has been assessed in gradient mode for the separation of phenol derivatives using columns packed with 2.6μm core-shell particles. The effect of mobile-phase velocity on peak
D Essaid et al.
Journal of pharmaceutical and biomedical analysis, 114, 227-240 (2015-06-24)
Porphyrins are a class of photosensitizers used in photodynamic therapy (PDT). Understanding the interaction of porphyrins with membrane cells components is important in order to improve this therapy. Many analytical methods can be used for this aim. High performance liquid
Martin Gilar et al.
Journal of chromatography. A, 1390, 86-94 (2015-03-10)
Chromatographic zone broadening is a common issue in microfluidic chromatography, where the sample volume introduced on column often exceeds the column void volume. To better understand the propagation of wide chromatographic zones on a separation device, a series of MS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service