Skip to Content
Merck
All Photos(2)

Key Documents

V900580

Sigma-Aldrich

N-Acetyl-L-glutamic acid

Vetec, reagent grade, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
189.17
Beilstein:
1727473
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

technique(s)

ligand binding assay: suitable

color

colorless to white

mp

194-196 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C7H11NO5/c1-4(9)8-5(7(12)13)2-3-6(10)11/h5H,2-3H2,1H3,(H,8,9)(H,10,11)(H,12,13)/t5-/m0/s1

InChI key

RFMMMVDNIPUKGG-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Zhang et al.
Clinical and investigative medicine. Medecine clinique et experimentale, 13(4), 183-188 (1990-08-01)
Liver tissue from 3 patients with carbamyl phosphate synthetase I deficiency and 11 patients with ornithine transcarbamylase deficiency was analyzed by Western blots for the presence of carbamyl phosphate synthetase I and ornithine transcarbamylase cross-reactive material with anti-rat enzyme antibodies.
M Tuchman et al.
Pediatric research, 27(4 Pt 1), 408-412 (1990-04-01)
N-acetylglutamate (NAG) content was measured in homogenates of liver and small intestine obtained from normal and 24-h starved syngeneic mice. Subsequently, NAG was determined in normal, and in carbamyl phosphate synthetase I and ornithine transcarbamylase enzyme-deficient human liver tissue homogenates.
J Vockley et al.
Biochemical medicine and metabolic biology, 47(1), 38-46 (1992-02-01)
N-Acetyl-L-glutamate synthetase (NAG synthetase) is a mitochondrial matrix enzyme which catalyzes the synthesis of N-acetyl-Lglutamate (NAG), a physiologic activator of the urea cycle enzyme carbamylphosphate synthetase I. Deficiency of NAG synthetase in humans has been reported only three times previously.
K Sugahara et al.
Journal of chromatography. B, Biomedical applications, 657(1), 15-21 (1994-07-01)
Liquid chromatography-atmospheric pressure chemical-ionization mass spectrometry (LC-APCI-MS) was used for the analysis of N-acetylamino acids that could not be determined with an amino acid analyzer. LC-APCI-MS could directly detect the protonated molecular ions of various synthetic N-acetylamino acids, distinguishing N-acetylserine
C M Puah et al.
Urological research, 10(2), 81-84 (1982-01-01)
A sensitive and reliable radioimmunoassay (RIA) for urinary unconjugated 5 alpha-androstane-3 alpha, 17 beta-diol is described. The mean overall recovery of unconjugated 5 alpha-androstane-3 alpha, 17 beta-diol was found to be 57.4%. The sensitivity of the assay was 79 fmol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service