Skip to Content
Merck
All Photos(3)

Key Documents

V900576

Sigma-Aldrich

1,8-Dihydroxyanthraquinone

Vetec, reagent grade, 96%

Synonym(s):

Chrysazine, Danthron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O4
CAS Number:
Molecular Weight:
240.21
Beilstein:
2054727
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

96%

mp

191-193 °C (lit.)

SMILES string

Oc1cccc2C(=O)c3cccc(O)c3C(=O)c12

InChI

1S/C14H8O4/c15-9-5-1-3-7-11(9)14(18)12-8(13(7)17)4-2-6-10(12)16/h1-6,15-16H

InChI key

QBPFLULOKWLNNW-UHFFFAOYSA-N

Gene Information

human ... RXRA(6256)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunyu Qiao et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 70(1), 136-143 (2007-09-11)
The interactions of fish sperm deoxyribonucleic acid (DNA) with anthraquinones, such as chrysophanol, physcion and 1,8-dihydroxy anthraquinone, were investigated by using ethidium bromide (EB) as fluorescence probe. The binding constants of anthraquinones and DNA were obtained by the fluorescence quenching
Hui Zhou et al.
Biophysical chemistry, 114(1), 21-26 (2005-03-29)
Danthron is an important natural occurring component in laxative drugs. In this paper, electrochemical investigation of danthron and its interaction with DNA is reported. Via the electrochemical approach assisted by ultraviolet-visible (UV-Vis) spectroscopy, we have proved that danthron intercalates into
Florian Pankewitz et al.
Journal of chemical ecology, 32(9), 2067-2072 (2006-07-13)
Eggs of several insect species are protected against natural enemies by noxious components. However, almost nothing is known about the fate of these defensive substances during egg development nor their site of biosynthesis. The eggs of several leaf beetle species
Hiroyuki Yamazaki et al.
Marine drugs, 10(12), 2691-2697 (2013-01-25)
1-Hydroxy-10-methoxy-dibenz[b,e]oxepin-6,11-dione (1) was obtained from the culture broth of a marine-derived fungus, Beauveria bassiana TPU942, isolated from a marine sponge collected at Iriomote Island in Okinawa, together with two known compounds, chrysazin (2) and globosuxanthone A (3). The structure of
Hsu-Feng Lu et al.
Neurochemical research, 35(7), 1105-1110 (2010-04-07)
Our primary studies had shown that danthron induced cytotoxic effects, including apoptosis and inhibition of migration and invasion. However, danthron-affected DNA damage and repair gene expressions are not clear. In this study, we investigated to examine whether or not danthron

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service