V900407
L-Aspartic acid
98.5-101.0%, Vetec™
Synonym(s):
(S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
HO2CCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
133.10
Beilstein:
1723530
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Product Name
L-Aspartic acid, Vetec™, reagent grade
grade
reagent grade
product line
Vetec™
Assay
98.5-101.0%
form
crystalline powder
color
white to off-white
mp
>300 °C (dec.) (lit.)
SMILES string
N[C@@H](CC(O)=O)C(O)=O
InChI
1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1
InChI key
CKLJMWTZIZZHCS-REOHCLBHSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Principal neurotransmitter for fast synaptic excitation.
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tamami Uejima et al.
Acta crystallographica. Section D, Biological crystallography, 69(Pt 3), 345-351 (2013-03-23)
Rab small GTPases regulate vesicle transport in eukaryotes by interacting with various effectors. Guanine nucleotide-exchange factor (GEF) catalyzes the transition from inactive GDP-bound Rab to active GTP-bound Rab. The existence of several GDP-bound intermediates containing the Arabidopsis thaliana Rab5 homologue
Guus B Erkens et al.
Nature, 502(7469), 119-123 (2013-10-05)
Excitatory amino acid transporters (EAATs) are secondary transport proteins that mediate the uptake of glutamate and other amino acids. EAATs fulfil an important role in neuronal signal transmission by clearing the excitatory neurotransmitters from the synaptic cleft after depolarization of
Ann-Louise Johansson et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(22), 8912-8917 (2013-05-16)
Proton transfer across biological membranes underpins central processes in biological systems, such as energy conservation and transport of ions and molecules. In the membrane proteins involved in these processes, proton transfer takes place through specific pathways connecting the two sides
T Kajikawa et al.
Journal of dental research, 93(4), 400-405 (2014-01-24)
PLAP-1/asporin is an extracellular matrix protein that is predominantly expressed in the human periodontal ligament (PDL) and has an aspartic acid (D) repeat polymorphism in its N-terminal region. In this study, we hypothesized that the D repeat polymorphism of PLAP-1/asporin
Yoshikatsu Kanai et al.
Molecular aspects of medicine, 34(2-3), 108-120 (2013-03-20)
Glutamate transporters play important roles in the termination of excitatory neurotransmission and in providing cells throughout the body with glutamate for metabolic purposes. The high-affinity glutamate transporters EAAC1 (SLC1A1), GLT1 (SLC1A2), GLAST (SLC1A3), EAAT4 (SLC1A6), and EAAT5 (SLC1A7) mediate the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service