Skip to Content
Merck
All Photos(3)

Key Documents

V900044

Sigma-Aldrich

Calcium chloride hexahydrate

Vetec, reagent grade, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 6H2O
CAS Number:
Molecular Weight:
219.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

97%

mp

30 °C

density

1.71 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.6H2O/h;2*1H;6*1H2/q+2;;;;;;;;/p-2

InChI key

QHFQAJHNDKBRBO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mamoun A Gharaibeh et al.
Environmental science and pollution research international, 22(23), 18894-18905 (2015-07-25)
Irrigation with wastewater is a promising option to improve crop yields and to reduce pressure on freshwater sources. However, heavy metal concentrations in wastewater may cause health concerns. A greenhouse pot experiment was conducted in order to determine cadmium (Cd)
Katrien Maes et al.
Journal of chromatography. A, 1360, 217-228 (2014-08-26)
Obtaining maximal sensitivity of nano UHPLC-MS/MS methods is primordial to quantify picomolar concentrations of neuropeptides in microdialysis samples. Since aspecific adsorption of peptides to Eppendorf tubes, pipette tips and UHPLC vials is detrimental for method sensitivity, a strategy is presented
Tiago Sousa et al.
Journal of pharmaceutical sciences, 103(10), 3171-3175 (2014-08-06)
Azo-bonded prodrugs of 5-aminosalicylic acid (mesalazine)-sulfasalazine, balsalazide, and olsalazine, which are used in the treatment of ulcerative colitis, rely on colonic bacteria to cleave the azo bond and liberate the active drug in the large intestine. The aim of this
Jie Wang et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 78, 31-36 (2015-06-27)
This study was the first to investigate the colonic stability of 17 peptide molecules (insulin, calcitonin, glucagon, secretin, somatostatin, desmopressin, oxytocin, Arg-vasopressin, octreotide, ciclosporin, leuprolide, nafarelin, buserelin, histrelin, [D-Ser(4)]-gonadorelin, deslorelin, and goserelin) in a model of the large intestine using
Sandeep Deshmukh et al.
Journal of chromatography. A, 1388, 1-8 (2015-03-01)
Accurate quantitation of compounds in samples of biological origin is often hampered by matrix interferences one of which occurs in GC-MS analysis from the presence of highly abundant phosphate. Consequently, high concentrations of phosphate need to be removed before sample

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service