V900030
Aluminum chloride hexahydrate
Vetec™, reagent grade, 98%
About This Item
grade
reagent grade
vapor pressure
1 mmHg ( 100 °C)
product line
Vetec™
Assay
98%
form
powder or crystals
reaction suitability
reagent type: catalyst
core: aluminum
pH
2.5-3.5 (20 °C)
anion traces
sulfate (SO42-): ≤100 ppm
cation traces
Fe: ≤100 ppm
Pb: ≤50 ppm
SMILES string
Cl[Al](Cl)Cl.O.O.O.O.O.O
InChI
1S/Al.3ClH.6H2O/h;3*1H;6*1H2/q+3;;;;;;;;;/p-3
InChI key
JGDITNMASUZKPW-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B - STOT RE 1
Target Organs
Lungs
Supplementary Hazards
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service