Skip to Content
Merck
All Photos(1)

Key Documents

V800058

Sigma-Aldrich

Ammonium thiosulfate

LR, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2S2O3
CAS Number:
Molecular Weight:
148.21
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
LR
form:
crystalline powder

grade

LR

product line

Vetec

Assay

≥98%

form

crystalline powder

density

1.679 g/mL at 25 °C (lit.)

anion traces

sulfate, sulfite (as SO42-): ≤1.00%

cation traces

Fe: ≤0.002%
heavy metals (as Pb): ≤0.002%

SMILES string

N.N.OS(O)(=O)=S

InChI

1S/2H3N.H2O3S2/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

XYXNTHIYBIDHGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Primary toxicological evaluation of ammonium thiosulfate].
Iu N Talakin et al.
Gigiena truda i professional'nye zabolevaniia, (6)(6), 54-55 (1982-06-01)
Ruijun Qin et al.
Journal of agricultural and food chemistry, 55(20), 8193-8199 (2007-09-13)
Low permeability tarps can effectively minimize fumigant emissions while improving fumigation efficacy by retaining fumigants under the tarp. However, when planting holes are cut through the tarps, high-concentration fumigants may be released and result in environmental and worker safety hazards.
L Cassina et al.
Journal of hazardous materials, 231-232, 36-42 (2012-07-10)
Mercury-contaminated soils from a petrochemical plant in southern Italy were investigated to assess the phytoextraction efficiency of crop plants treated with the phytohormone, cytokinine (CK foliar treatment), and with the thioligand, ammonium thiosulfate (TS, soil application). Plant biomass, evapotranspiration, Hg
Bikshandarkoil R Srinivasan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 102, 235-241 (2012-12-12)
Aqueous reaction of ammonium thiosulfate with ethylenediamine (en) results in the formation of the title compound (enH(2))[S(2)O(3)] (1) (enH(2)=ethylenediammonium) in good yields. Compound 1 was characterized by analytical data, IR, Raman and NMR spectra, X-ray powder pattern and its crystal
Fabio N Moreno et al.
Environmental pollution (Barking, Essex : 1987), 136(2), 341-352 (2005-04-21)
Experiments were carried out in plant growth chambers and in the field to investigate plant-mercury accumulation and volatilisation in the presence of thiosulphate (S2O3)-containing solutions. Brassica juncea (Indian mustard) plants grown in Hg-contaminated Tui mine tailings (New Zealand) were enclosed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service