Skip to Content
Merck
All Photos(1)

Key Documents

V001811

Sigma-Aldrich

Alizarin Yellow GG

Synonym(s):

5-(3-Nitrophenylazo)salicylic acid sodium salt, Alizarin Yellow 2G, Metachrome Yellow, Mordant Yellow 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8N3NaO5
CAS Number:
Molecular Weight:
309.21
Beilstein:
3820455
EC Number:
MDL number:
PubChem Substance ID:

SMILES string

[Na+].Oc1ccc(cc1C([O-])=O)\N=N\c2cccc(c2)[N+]([O-])=O

InChI

1S/C13H9N3O5.Na/c17-12-5-4-9(7-11(12)13(18)19)15-14-8-2-1-3-10(6-8)16(20)21;/h1-7,17H,(H,18,19);/q;+1/p-1/b15-14+;

InChI key

ZHFPEICFUVWJIS-WPDLWGESSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service