Skip to Content
Merck
All Photos(1)

Documents

1379276

USP

Melengestrol Acetate Related B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

6,16-Dimethylene-17-hydroxypregna-4-ene-3,20-dione 17-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32O4
CAS Number:
Molecular Weight:
396.52
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

melengestrol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C25H32O4/c1-14-11-19-20(23(5)9-7-18(28)13-21(14)23)8-10-24(6)22(19)12-15(2)25(24,16(3)26)29-17(4)27/h13,19-20,22H,1-2,7-12H2,3-6H3/t19-,20+,22+,23-,24+,25+/m1/s1

InChI key

WJFDQNDYQVFWKP-DWNQPYOZSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Melengestrol Acetate Related B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Melengestrol Acetate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service