Skip to Content
Merck
All Photos(1)

Key Documents

ULSD20-100

ULTRA LOW SULFUR IN DIESEL OIL, 0-100PPM

set of 5 × 100 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

packaging

set of 5 × 100 mL

concentration

20-100 ppm

storage temp.

room temp

General description

Sulfur with Ultra Low Diesel Fuel. Concentrations of 20, 40, 60, 80 and 100 ppm. 100 mL each.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service