PHR9051
DL-Kynurenine
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(2RS)-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid, 2-Amino-3-(2-aminobenzoyl)propionic acid, 2-Amino-4-(2-aminophenyl)-4-oxobutyric acid
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
API family
kynurenic acid
product line
TraceCERT®
form
solid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
mp
~235 °C (dec.)
application(s)
pharmaceutical small molecule
SMILES string
NC(CC(=O)c1ccccc1N)C(O)=O
InChI
1S/C10H12N2O3/c11-7-4-2-1-3-6(7)9(13)5-8(12)10(14)15/h1-4,8H,5,11-12H2,(H,14,15)
InChI key
YGPSJZOEDVAXAB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com
Application
Other Notes
Recommended products
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service