Skip to Content
Merck
All Photos(1)

Documents

PHL80087

DL-Camphor

phyproof® Reference Substance

Synonym(s):

(±)-Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
1907611
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

vapor density

5.2 (vs air)

vapor pressure

4 mmHg ( 70 °C)

product line

phyproof® Reference Substance

Assay

≥90.0% (GC)

form

powder

expl. lim.

3.5 %

manufacturer/tradename

PhytoLab

bp

204 °C (lit.)

mp

175-177 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][C@](CC1=O)(CC2)C(C)(C)[C@]12C

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: cinnamomum curcuma salvia

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.9 °F - closed cup

Flash Point(C)

64.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service