G005961
HPLC Separation of an Oligonucleotide and Impurities on Proteomix® SAX-NP5 versus Porous SAX
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: Proteomix SAX-NP5, 15 cm x 4.6 mm I.D., 5 μm particles (Z777228)
mobile phase: (A) 0.025 M Tris buffer pH9; (B) A + 1.0 M LiCi
gradient: 30 to 75% B in 45 min
flow rate: 1.0 mL/min
detector: UV, 260 nm
injection: 50 μL
sample: 50-mer Oligo
suitability
application for HPLC
application(s)
forensics and toxicology
life science and biopharma
Legal Information
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service