G005921
GC Analysis of PCBs in Whole Cow Milk (Spiked) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis
suitable for GC, application for SPE
About This Item
Recommended Products
technique(s)
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable
test parameters
sample preparation: SPE (Solid Phase Extraction)
sample/matrix: 10 g whole cow′s milk
extraction tube: Supel™ QuE Non-buffered tube (4 g magnesium sulfate, 1 g sodium chloride) (55294-U)
clean-up tube: Supel™ QuE Z-Sep 150 mg, magnesium sulfate (150 mg, part no. 63135) (55299-U)
column: SLB-5ms, 20 m x 0.18 mm I.D., 0.18 μm (28564-U)
oven: 75 °C (1 min), 12 °C/min to 340 °C (10 min)
inj. temp.: 250 °C
detector: μ-ECD, 340 °C
carrier gas: hydrogen, 1.2 mL/min, constant
injection: 1 μL splitless (0.75 min)
liner: 4 mm I.D. FocusLiner™ with taper
suitability
application for SPE
application(s)
food and beverages
General description
Analysis Note
Extraction solvent: acetone:hexane (1:1), 20 mL
Clean-up volume: 1 mL
Legal Information
SPE tube or plate
analytical column
standard
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service