G003779
GC Analysis of Semivolatiles on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
application for SPME, suitable for GC
About This Item
technique(s)
gas chromatography (GC): suitable
solid phase microextraction (SPME): suitable
test parameters
sample/matrix: Base neutral analytes at 50 ppb and internal standards 25 ppb in 0.1M phosphate buffer, pH 11, with 25% NaCl, 9 mL in 10 mL screw cap vial
SPME fiber: 60 μm Carbowax (PEG) (57354-U)
extraction: 30 min. direct immersion with agitation using CombiPal autosampler
desorption process: 2 min at 250°C, splitless for 0.75 min then opened
column: SLB-5ms, 30 m x 0.25 mm I.D., 0.5μm film
oven: 50°C(2 min) to 130°C @ 12°C/min to 200°C @ 20°C/min to 260°C @ 15°C/min to 310°C @ 20°C/min (4 min)
inj. temp.: Splitless/split with 0.75mm ID liner and Merlin Microseal
detector: ITMS m/z=50-230 @ 0.65 μsec per scan, transfer line 310°C
carrier gas: Helium, 1mL/min constant flow, 8.7 psi
suitability
application for SPME
application(s)
environmental
food and beverages
Legal Information
Analyte
SPME fiber
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service