Skip to Content
Merck
All Photos(1)

Key Documents

G003775

GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

application for SPME, suitable for GC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable
solid phase microextraction (SPME): suitable

test parameters

sample/matrix: Pesticides at 10 ppb in 0.1M phosphate buffer, pH 7, with 25% NaCl, 9 mL in 10 mL screw cap vial
SPME fiber: 60 μm Carbowax (PEG) (57354-U)
extraction: 30 min. direct immersion with agitation using CombiPal autosampler
column: SLB-5ms, 30 m x 0.25 mm I.D., 0.5μm film
desorption process: 2 min. at 250°C, splitless for 0.75 min. then opened
oven: 60°C(1 min) to 150°C @ 25°C/min to 230°C @ 5°C/min to 310°C @ 18°C/min (3 min)
inj. temp.: Splitless/split with 0.75mm ID liner and Merlin Microseal
detector: ITMS m/z=60-450 @ 0.9 μsec per scan, transfer line 310°C
carrier gas: Helium, 1mL/min constant flow

suitability

application for SPME

application(s)

agriculture
chemicals and industrial polymers
environmental
food and beverages

Legal Information

Carbowax is a registered trademark of Union Carbide
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Triethyl thiophosphate
    Thionazin
    Phorate
    α-1,2,3,4,5,6-Hexachlorocyclohexane
    Dimethoate
    Prometon
    Simazine
    Atrazine
    Propazine
    β-BHC
    Terbufos
    δ-BHC
    Diazinon
    Disulfoton
    γ-BHC, certified reference material, 1000 μg/mL in methanol
    Methyl parathion, analytical standard
    Simetryn
    Ametryn
    Prometryn
    Heptachlor
    Terbutryn
    Malathion
    Cyanazine
    Parathion
    Aldrin
    Merphos, PESTANAL®, analytical standard
    Fenamiphos
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene
    Dieldrin
    Endrin
    Endrin aldehyde
    4,4′-DDT
    Endosulfan sulfate
    Methoxychlor
    Endrin ketone
    See All (35)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service