Skip to Content
Merck
All Photos(1)

Key Documents

CRM46286

Supelco

Flurtamone

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

5-(Methylamino)-2-phenyl-4-[3-(trifluoromethyl)phenyl]-3(2H)-furanone

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€76.50

€76.50


Please contact Customer Service for Availability


Select a Size

Change View
50 MG
€76.50

About This Item

Empirical Formula (Hill Notation):
C18H14F3NO2
CAS Number:
Molecular Weight:
333.30
Beilstein:
7543933
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€76.50


Please contact Customer Service for Availability

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

Limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

CNC1=C(C(=O)C(O1)c2ccccc2)c3cccc(c3)C(F)(F)F

InChI

1S/C18H14F3NO2/c1-22-17-14(12-8-5-9-13(10-12)18(19,20)21)15(23)16(24-17)11-6-3-2-4-7-11/h2-10,16,22H,1H3

InChI key

NYRMIJKDBAQCHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Application

Flurtamone is a pre-emergence or post-emergence herbicide used pre-planting and soil incroporated to control broad-leaved weeds and grasses such as barley, wheat, oats, rye, sunflower, cotton, peas. Flurtamone is not approved for use in the European Union as a plant protection product. Maximum residue levels (MRLs) are set according to Reg. (EU) 2016/1016 for products of plant and animal origin in the range 0.01 - 0.05 mg/kg . The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service