Skip to Content
Merck
All Photos(1)

Documents

CRM03903

Supelco

Resmethrin

mixture of stereoisomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26O3
CAS Number:
Molecular Weight:
338.44
Beilstein:
1626510
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

description

mixture of isomers

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

C\C(C)=C\C1C(C(=O)OCc2coc(Cc3ccccc3)c2)C1(C)C

InChI

1S/C22H26O3/c1-15(2)10-19-20(22(19,3)4)21(23)25-14-17-12-18(24-13-17)11-16-8-6-5-7-9-16/h5-10,12-13,19-20H,11,14H2,1-4H3

InChI key

VEMKTZHHVJILDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034.. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com


Resmethrin is a synthetic pyrethroid insecticide with insecticidal activity similar to natural pyrethrin. It is used for the protection of crops and food storage against insects and acarids.


As per Regulation (EC) No 1107/2009, repealing the directive 91/414/ECC, resmethrin is not approved for use in the European Union(EU). A maximum residue limit (MRL) of 0.01 mg/kg is applicable for the presence of resmethrin (including other mixtures of constituent isomers (sum of isomers)) in fresh/frozen fruits, and tree nuts, in accordance with Regulation (EU) 2015/868.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.The certified reference material resmethrin may find the following uses:
  • To evaluate the toxicity of resmethrin on the male reproductive system using a testis organ-culture method
    Quantification of tetramethrin and resmethrin in human body fluids using liquid chromatography (LC)–tandem mass spectrometry (MS/MS)
  • Characterization of the interaction between resmethrin and calf thymus DNA in vitro by multiple spectroscopic methods
  • Heterogeneous reactions study of resmethrin, phenothrin, and fenvalerate particles with nitrate radicals
  • Synthetic pyrethroid pesticides toxicity in Caenorhabditis elegans to analyze physiological change

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service